Gene Transcripts

Gene Symbol
ASL
Entrez Gene ID
435
Gene Name
argininosuccinate lyase
Gene Aliases
ASAL
Chromosome Location
Chr. 7: 66075789 - 66093343 on Build GRCh38
UniGene ID
Hs.632015
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000048.3 NP_000039.2 4 352
NM_001024943.1 NP_001020114.1 3 476
NM_001024944.1 NP_001020115.1 3 476
NM_001024946.1 NP_001020117.1 3 476
GenBank mRNA AK300519.1 - 4 307
AK310912.1 - 3 482
AK316334.1 - 3 372
AY203938.1 - 3 1351
BC008195.1 - 4 314 3048146
BC033146.1 - 3 287 4101398
BQ949510.1 - 3 476
BX328836.2 - 4 372
J03058.1 - 4 310
KU177920.1 - 2 241
KU177921.1 - 2 241
KU177922.1 - 2 241
M14218.1 - 3 355
M57638.1 - 3 286
Y00753.1 - 3 286

View Full Product Details