Gene Transcripts

Gene Symbol
Pstk
Entrez Gene ID
361661
Gene Name
phosphoseryl-tRNA kinase
Gene Aliases
RGD1564300
Chromosome Location
Chr. 1: 201950148 - 201960563 on Build Rnor_6.0
UniGene ID
Rn.199179
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271229.1 NP_001258158.1 6 1045
GenBank mRNA BC168162.1 - 5 1279

View Full Product Details