Gene Transcripts

Gene Symbol
Tsga10ip
Entrez Gene ID
361707
Gene Name
testis specific 10 interacting protein
Gene Aliases
Chromosome Location
Chr. 1: 220772592 - 220786931 on Build Rnor_6.0
UniGene ID
Rn.209597
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004278.3 NP_001004278.2 4, 5 1158
GenBank mRNA AY690667.1 - 4, 5 1175

View Full Product Details