Gene Transcripts

Gene Symbol
Zdhhc6
Entrez Gene ID
361771
Gene Name
zinc finger, DHHC-type containing 6
Gene Aliases
Chromosome Location
Chr. 1: 276291411 - 276309633 on Build Rnor_6.0
UniGene ID
Rn.19409
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037652.1 NP_001032741.1 546
GenBank mRNA AY886524.1 - 1 427
BC107928.1 - 546 7130799

View Full Product Details