Gene Transcripts
- Gene Symbol
- Ints3
- Entrez Gene ID
- 361988
- Gene Name
- integrator complex subunit 3
- Gene Aliases
- RGD1304593
- Chromosome Location
- Chr. 2: 189766927 - 189818322 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_342288.6 |
XP_342289.2
|
13 |
2049 |
|
View Full Product Details