Gene Transcripts

Gene Symbol
Dnajb6
Entrez Gene ID
362293
Gene Name
DnaJ (Hsp40) homolog, subfamily B, member 6
Gene Aliases
Chromosome Location
Chr. 4: 2711339 - 2774969 on Build Rnor_6.0
UniGene ID
Rn.129050
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013209.1 NP_001013227.1 3 251
XM_006235863.2 XP_006235925.1 3 297
XM_006235864.2 XP_006235926.1 4 354
XM_006235866.2 XP_006235928.1 3 294
GenBank mRNA BC078908.1 - 3 251
FQ214601.1 - 3 252

Gene Symbol
LOC690183
Entrez Gene ID
690183
Gene Name
similar to DnaJ homolog subfamily B member 6 (Heat shock protein J2) (HSJ-2) (MRJ) (mDj4)
Gene Aliases
Chromosome Location
Chr. 1: 223548728 - 223550015 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001073579.4 XP_001073579.1 1 245

View Full Product Details