Gene Transcripts

Gene Symbol
Clec4a1
Entrez Gene ID
362430
Gene Name
C-type lectin domain family 4, member A1
Gene Aliases
Dcir4
Chromosome Location
Chr. 4: 155947794 - 155959909 on Build Rnor_6.0
UniGene ID
Rn.207391
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005890.2 NP_001005890.1 2, 3 322
GenBank mRNA AY397672.1 - 2, 3 223
AY494063.1 - 2, 3 223
BC098830.1 - 2, 3 322 7378545

View Full Product Details