Gene Transcripts

Gene Symbol
Plekhf2
Entrez Gene ID
362484
Gene Name
pleckstrin homology domain containing, family F (with FYVE domain) member 2
Gene Aliases
Chromosome Location
Chr. 5: 24240241 - 24257099 on Build Rnor_6.0
UniGene ID
Rn.11991
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108655.1 NP_001102125.1 2 509
XM_006237872.2 XP_006237934.1 2 2017
GenBank mRNA BC161902.1 - 2 498

View Full Product Details