Gene Transcripts

Gene Symbol
Pate4
Entrez Gene ID
363041
Gene Name
prostate and testis expressed 4
Gene Aliases
Svs7
Chromosome Location
Chr. 8: 36815005 - 36817747 on Build Rnor_6.0
UniGene ID
Rn.78071
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108757.1 NP_001102227.1 2 185
XM_008766061.1 XP_008764283.1 1 319

View Full Product Details