Gene Transcripts

Gene Symbol
Mterfd2
Entrez Gene ID
363289
Gene Name
MTERF domain containing 2
Gene Aliases
MTERF4
Chromosome Location
Chr. 9: 100440482 - 100445158 on Build Rnor_6.0
UniGene ID
Rn.41249
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037209.1 NP_001032286.1 2 81
GenBank mRNA BC098676.1 - 2 81

View Full Product Details