Gene Transcripts

Gene Symbol
Cyb5d1
Entrez Gene ID
363629
Gene Name
cytochrome b5 domain containing 1
Gene Aliases
RGD1559567
Chromosome Location
Chr. 10: 55993206 - 55997426 on Build Rnor_6.0
UniGene ID
Rn.208132
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191890.1 NP_001178819.1 1 147
XM_006246767.2 XP_006246829.1 2 199
GenBank mRNA FQ223142.1 - 1 538

View Full Product Details