Gene Transcripts

Gene Symbol
Sp6
Entrez Gene ID
363672
Gene Name
Sp6 transcription factor
Gene Aliases
Chromosome Location
Chr. 10: 84955864 - 84960045 on Build Rnor_6.0
UniGene ID
Rn.218646
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108833.1 NP_001102303.1 2 872

View Full Product Details