Gene Transcripts

Gene Symbol
Il21
Entrez Gene ID
365769
Gene Name
interleukin 21
Gene Aliases
IL-21
Chromosome Location
Chr. 2: 123965021 - 123972356 on Build Rnor_6.0
UniGene ID
Rn.214949
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108943.2 NP_001102413.1 3 247
GenBank mRNA DQ387062.1 - 3 247

View Full Product Details