Gene Transcripts

Gene Symbol
Crb2
Entrez Gene ID
366031
Gene Name
crumbs family member 2
Gene Aliases
RGD1309368
Chromosome Location
Chr. 3: 22038112 - 22061247 on Build Rnor_6.0
UniGene ID
Rn.195474
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135761.1 NP_001129233.1 10 3284
XM_006234103.2 XP_006234165.1 9 3415

View Full Product Details