Gene Transcripts

Gene Symbol
Ptch2
Entrez Gene ID
366452
Gene Name
patched 2
Gene Aliases
Ptch1
Chromosome Location
Chr. 5: 135962252 - 135984730 on Build Rnor_6.0
UniGene ID
Rn.105585
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108975.1 NP_001102445.1 16 2466
XM_008764050.1 XP_008762272.1 16 3351

View Full Product Details