Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278612.1 | NP_001265541.1 | 5 | 500 | |
NM_004681.3 | NP_004672.2 | 6 | 551 | ||
GenBank mRNA | AF000987.1 | - | 6 | 522 | |
AK127664.1 | - | 1 | 2819 | ||
BC005248.1 | - | 6 | 433 | 3933718 | |
BP331805.1 | - | 6 | 551 | ||
BT007201.1 | - | 6 | 390 | ||
DQ786267.1 | - | 5 | 423 |