Gene Transcripts

Gene Symbol
Mrgprh
Entrez Gene ID
404641
Gene Name
MAS-related GPR, member H
Gene Aliases
Mgrb, MrgHGPR90
Chromosome Location
Chr. 1: 48068406 - 48076715 on Build Rnor_6.0
UniGene ID
Rn.142013
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002281.1 NP_001002281.1 1 799
XM_006227882.2 XP_006227944.1 2 1051

View Full Product Details