Gene Transcripts

Gene Symbol
Clec4b2
Entrez Gene ID
450222
Gene Name
C-type lectin domain family 4, member B2
Gene Aliases
Aplra1
Chromosome Location
Chr. 4: 156107720 - 156131226 on Build Rnor_6.0
UniGene ID
Rn.129075
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005896.1 NP_001005896.1 4 312
GenBank mRNA AY365132.1 - 4 312
AY494066.1 - 4 279
AY581050.1 - 4 279

View Full Product Details