Gene Transcripts
- Gene Symbol
- Sowaha
- Entrez Gene ID
- 497903
- Gene Name
- sosondowah ankyrin repeat domain family member A
- Gene Aliases
- Ankrd43, RGD1564616
- Chromosome Location
- Chr. 10: 38836158 - 38838917 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_573089.5 |
XP_573089.1
|
1 |
829 |
|
View Full Product Details