Gene Transcripts

Gene Symbol
Commd6
Entrez Gene ID
498559
Gene Name
COMM domain containing 6
Gene Aliases
RGD1560745
Chromosome Location
Chr. 15: 86136096 - 86142672 on Build Rnor_6.0
UniGene ID
Rn.40372
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109105.1 NP_001102575.1 4 731
GenBank mRNA FQ223060.1 - 4 531
FQ224504.1 - 2 380

View Full Product Details