Gene Transcripts

Gene Symbol
RGD1560020_predicted
Entrez Gene ID
498982
Gene Name
similar to Myb proto-oncogene protein (C-myb) (predicted)
Gene Aliases
Chromosome Location
Chr. 1: 16658178 - 16690135 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008758639.1 XP_008756861.1 12 4136

View Full Product Details