Gene Transcripts

Gene Symbol
Naaladl2
Entrez Gene ID
499583
Gene Name
N-acetylated alpha-linked acidic dipeptidase-like 2
Gene Aliases
RGD1565147
Chromosome Location
Chr. 2: 109092931 - 110424878 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760899.1 XP_008759121.1 4 2385

View Full Product Details