Gene Transcripts

Gene Symbol
Duoxa2
Entrez Gene ID
499879
Gene Name
dual oxidase maturation factor 2
Gene Aliases
RGD1560628
Chromosome Location
Chr. 3: 114236718 - 114240086 on Build Rnor_6.0
UniGene ID
Rn.228195
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191965.1 NP_001178894.1 2 187

View Full Product Details