Gene Transcripts

Gene Symbol
NCR2
Entrez Gene ID
9436
Gene Name
natural cytotoxicity triggering receptor 2
Gene Aliases
CD336, LY95, NK-p44, NKP44, dJ149M18.1
Chromosome Location
Chr. 6: 41335620 - 41350887 on Build GRCh38
UniGene ID
Hs.194721
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199509.1 NP_001186438.1 2 325
NM_001199510.1 NP_001186439.1 2 325
NM_004828.3 NP_004819.2 2 325
XM_017011500.1 XP_016866989.1 2 495
GenBank mRNA AJ010099.1 - 2 264
AJ010100.1 - 2 264

View Full Product Details