Gene Transcripts

Gene Symbol
Vom2r36
Entrez Gene ID
502301
Gene Name
vomeronasal 2 receptor, 36
Gene Aliases
RGD1560365
Chromosome Location
Chr. 1: 65906344 - 66159639 on Build Rnor_6.0
UniGene ID
Rn.217936
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099483.1 NP_001092953.1 2 596

View Full Product Details