Gene Transcripts

Gene Symbol
Nkx3-2
Entrez Gene ID
682329
Gene Name
NK3 homeobox 2
Gene Aliases
Chromosome Location
Chr. 14: 73813531 - 73817888 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008770239.1 XP_008768461.1 2 746

View Full Product Details