Gene Transcripts

Gene Symbol
Ecscr
Entrez Gene ID
689248
Gene Name
endothelial cell surface expressed chemotaxis and apoptosis regulator
Gene Aliases
Aria, Ecscr-ps1
Chromosome Location
Chr. 18: 28507167 - 28516237 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008772057.1 XP_008770279.1 8 660
XM_008772058.1 XP_008770280.1 7 615

View Full Product Details