Gene Transcripts
- Gene Symbol
- RALGPS1
- Entrez Gene ID
- 9649
- Gene Name
- Ral GEF with PH domain and SH3 binding motif 1
- Gene Aliases
- RALGEF2, RALGPS1A
- Chromosome Location
- Chr. 9: 126914774 - 127223166 on Build GRCh38
- UniGene ID
- Hs.432842
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001190728.1 |
NP_001177657.1
|
6, 7 |
553 |
|
NM_001190729.1 |
NP_001177658.1
|
7, 8 |
792 |
|
NM_001190730.1 |
NP_001177659.1
|
7, 8 |
792 |
|
NM_001322320.1 |
NP_001309249.1
|
7, 8 |
755 |
|
NM_001322321.1 |
NP_001309250.1
|
7, 8 |
792 |
|
NM_001322322.1 |
NP_001309251.1
|
7, 8 |
792 |
|
NM_001322323.1 |
NP_001309252.1
|
6, 7 |
702 |
|
NM_001322324.1 |
NP_001309253.1
|
7, 8 |
792 |
|
NM_001322325.1 |
NP_001309254.1
|
7, 8 |
792 |
|
NM_014636.2 |
NP_055451.1
|
7, 8 |
755 |
|
XM_006717328.3 |
XP_006717391.2
|
7, 8 |
604 |
|
XM_011519225.1 |
XP_011517527.1
|
6, 7 |
553 |
|
XM_011519226.1 |
XP_011517528.1
|
6, 7 |
553 |
|
XM_011519227.1 |
XP_011517529.1
|
5, 6 |
502 |
|
XM_011519228.1 |
XP_011517530.1
|
6, 7 |
553 |
|
XM_011519229.1 |
XP_011517531.1
|
7, 8 |
610 |
|
XM_011519230.2 |
XP_011517532.1
|
7, 8 |
604 |
|
XM_011519231.1 |
XP_011517533.1
|
6, 7 |
553 |
|
XM_011519232.1 |
XP_011517534.1
|
6, 7 |
553 |
|
XM_011519233.1 |
XP_011517535.1
|
6, 7 |
553 |
|
XM_011519234.2 |
XP_011517536.1
|
6, 7 |
687 |
|
XM_011519235.2 |
XP_011517537.1
|
7, 8 |
778 |
|
XM_011519236.1 |
XP_011517538.1
|
6, 7 |
553 |
|
XM_011519238.2 |
XP_011517540.1
|
6, 7 |
553 |
|
XM_017015342.1 |
XP_016870831.1
|
6, 7 |
553 |
|
XM_017015343.1 |
XP_016870832.1
|
7, 8 |
597 |
|
XM_017015345.1 |
XP_016870834.1
|
6, 7 |
553 |
|
XM_017015346.1 |
XP_016870835.1
|
6, 7 |
553 |
|
XM_017015347.1 |
XP_016870836.1
|
7, 8 |
612 |
|
XM_017015348.1 |
XP_016870837.1
|
7, 8 |
786 |
|
XM_017015349.1 |
XP_016870838.1
|
3, 4 |
238 |
|
XM_017015350.1 |
XP_016870839.1
|
6, 7 |
553 |
|
GenBank mRNA |
AB002349.1 |
- |
7, 8 |
755 |
|
AF221098.1 |
- |
6, 7 |
553 |
|
AK299149.1 |
- |
7, 8 |
733 |
|
BC032372.1 |
- |
7, 8 |
715 |
5163332 |
BC033708.1 |
- |
7, 8 |
785 |
3901593 |
DR001330.1 |
- |
7, 8 |
747 |
|
View Full Product Details