Gene Transcripts

Gene Symbol
Muc5ac
Entrez Gene ID
682837
Gene Name
mucin 5AC, oligomeric mucus/gel-forming
Gene Aliases
Chromosome Location
Chr. 1: 214725485 - 214756653 on Build Rnor_6.0
UniGene ID
Rn.10789
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760037.1 XP_008758259.1 4 472
GenBank mRNA AB042530.1 - 4 496

View Full Product Details