Gene Transcripts

Gene Symbol
KIAA0100
Entrez Gene ID
9703
Gene Name
KIAA0100
Gene Aliases
BCOX, BCOX1, CT101, FMP27
Chromosome Location
Chr. 17: 28614440 - 28645159 on Build GRCh38
UniGene ID
Hs.151761
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014680.3 NP_055495.2 16 2602
XM_005258073.2 XP_005258130.1 16 2597
XM_017025423.1 XP_016880912.1 15 2451
GenBank mRNA BC120873.1 - 16 2461
CR933634.1 - 17 2464
D43947.3 - 16 2574

View Full Product Details