Gene Transcripts

Gene Symbol
Rab20
Entrez Gene ID
689377
Gene Name
RAB20, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 16: 83358116 - 83382326 on Build Rnor_6.0
UniGene ID
Rn.140755
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109535.1 NP_001103005.1 2 710

View Full Product Details