Gene Transcripts

Gene Symbol
Bcl3
Entrez Gene ID
680611
Gene Name
B-cell CLL/lymphoma 3
Gene Aliases
Chromosome Location
Chr. 1: 80730758 - 80745086 on Build Rnor_6.0
UniGene ID
Rn.198135
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109422.1 NP_001102892.1 3 519
XM_006228473.2 XP_006228535.1 3 612

View Full Product Details