Gene Transcripts

Gene Symbol
Pdf
Entrez Gene ID
690214
Gene Name
peptide deformylase (mitochondrial)
Gene Aliases
Chromosome Location
Chr. 19: 39244246 - 39246635 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001073696.5 XP_001073696.3 1 482

View Full Product Details