Gene Transcripts

Gene Symbol
Gpr81
Entrez Gene ID
689936
Gene Name
G protein-coupled receptor 81
Gene Aliases
Chromosome Location
Chr. 12: 38145177 - 38146232 on Build Rnor_6.0
UniGene ID
Rn.198299
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145334.1 NP_001138806.1 1 243
GenBank mRNA EU809461.1 - 1 243
EU809462.1 - 1 219

View Full Product Details