Gene Transcripts

Gene Symbol
Gypa
Entrez Gene ID
688972
Gene Name
glycophorin A
Gene Aliases
Chromosome Location
Chr. 19: 31115545 - 31131079 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001069014.4 XP_001069014.1 6 449
XM_006255409.2 XP_006255471.1 6 424

View Full Product Details