Gene Transcripts

Gene Symbol
Peli1
Entrez Gene ID
305549
Gene Name
pellino E3 ubiquitin protein ligase 1
Gene Aliases
Chromosome Location
Chr. 14: 106008086 - 106061641 on Build Rnor_6.0
UniGene ID
Rn.22814
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100565.1 NP_001094035.1 6 935
GenBank mRNA BC166745.1 - 6 768

View Full Product Details