Gene Transcripts

Gene Symbol
UTS2R
Entrez Gene ID
2837
Gene Name
urotensin 2 receptor
Gene Aliases
GPR14, UR-2-R, UTR, UTR2
Chromosome Location
Chr. 17: 82374277 - 82375586 on Build GRCh38
UniGene ID
Hs.192720
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018949.2 NP_061822.1 1 142
GenBank mRNA BC136490.1 - 1 142

View Full Product Details