Gene Transcripts

Gene Symbol
HIST1H3C
Entrez Gene ID
8352
Gene Name
histone cluster 1, H3c
Gene Aliases
H3.1, H3/c, H3FC
Chromosome Location
Chr. 6: 26045411 - 26045869 on Build GRCh38
UniGene ID
Hs.248176
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003531.2 NP_003522.1 1 412
GenBank mRNA BC127610.1 - 1 412

View Full Product Details