Gene Transcripts

Gene Symbol
RLN3
Entrez Gene ID
117579
Gene Name
relaxin 3
Gene Aliases
H3, RXN3, ZINS4, insl7
Chromosome Location
Chr. 19: 14028148 - 14031558 on Build GRCh38
UniGene ID
Hs.352155
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080864.3 NP_543140.1 1, 2 248
GenBank mRNA AB076563.1 - 1, 2 191
AF447451.1 - 1, 2 191
AY358181.1 - 2, 3 1031
BC140935.1 - 1, 2 248 9056747

View Full Product Details