Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_080752.3 | NP_542790.2 | 1 | 376 | |
NR_037628.1 | 1 | 376 | |||
GenBank mRNA | AK056641.1 | - | 1 | 341 | |
AK302767.1 | - | 1 | 340 | ||
AK316214.1 | - | 1 | 339 | ||
BC027611.2 | - | 1 | 333 | 4829980 | |
BX385024.2 | - | 1 | 334 | ||
DA520427.1 | - | 1 | 376 | ||
DC363945.1 | - | 1 | 339 |