Gene Transcripts

Gene Symbol
KLRG2
Entrez Gene ID
346689
Gene Name
killer cell lectin like receptor G2
Gene Aliases
CLEC15B
Chromosome Location
Chr. 7: 139426931 - 139483716 on Build GRCh38
UniGene ID
Hs.17572
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198508.2 NP_940910.1 5 1494
GenBank mRNA AK126174.1 - 5 1499
BC110858.1 - 2 1094 6139360

View Full Product Details