Gene Transcripts

Gene Symbol
KRTAP5-1
Entrez Gene ID
387264
Gene Name
keratin associated protein 5-1
Gene Aliases
KRN1L, KRTAP5.1
Chromosome Location
Chr. 11: 1584342 - 1585283 on Build GRCh38
UniGene ID
Hs.632147
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005922.1 NP_001005922.1 1 790
GenBank mRNA AB126070.1 - 1 790

View Full Product Details