Gene Transcripts

Gene Symbol
CT47A7
Entrez Gene ID
653282
Gene Name
cancer/testis antigen family 47, member A7
Gene Aliases
CT47.7
Chromosome Location
Chr. X: 120953282 - 120956600 on Build GRCh38
UniGene ID
Hs.712504
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080140.1 NP_001073609.1 3 1168

Gene Symbol
CT47A1
Entrez Gene ID
728096
Gene Name
cancer/testis antigen family 47, member A1
Gene Aliases
CT47.1
Chromosome Location
Chr. X: 120982466 - 120985886 on Build GRCh38
UniGene ID
Hs.729566
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080146.1 NP_001073615.1 3 1168

Gene Symbol
CT47A10
Entrez Gene ID
728036
Gene Name
cancer/testis antigen family 47, member A10
Gene Aliases
CT47, CT47.10
Chromosome Location
Chr. X: 120938701 - 120942121 on Build GRCh38
UniGene ID
Hs.729566
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080137.1 NP_001073606.1 3 1168

Gene Symbol
CT47A11
Entrez Gene ID
255313
Gene Name
cancer/testis antigen family 47, member A11
Gene Aliases
CT47.11
Chromosome Location
Chr. X: 120933840 - 120937260 on Build GRCh38
UniGene ID
Hs.522775
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173571.2 NP_775842.2 3 1168
GenBank mRNA BC029540.1 - 3 1172

Gene Symbol
CT47A12
Entrez Gene ID
100507170
Gene Name
cancer/testis antigen family 47, member A12
Gene Aliases
CT47, CT47.12
Chromosome Location
Chr. X: 120877490 - 120932399 on Build GRCh38
UniGene ID
Hs.729566
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242922.1 NP_001229851.1 3 1168

Gene Symbol
CT47A2
Entrez Gene ID
728090
Gene Name
cancer/testis antigen family 47, member A2
Gene Aliases
CT47.2
Chromosome Location
Chr. X: 120977606 - 120981026 on Build GRCh38
UniGene ID
Hs.729566
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080145.1 NP_001073614.1 3 1168

Gene Symbol
CT47A3
Entrez Gene ID
728082
Gene Name
cancer/testis antigen family 47, member A3
Gene Aliases
CT47.3
Chromosome Location
Chr. X: 120972746 - 120976166 on Build GRCh38
UniGene ID
Hs.729566
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080144.1 NP_001073613.1 3 1168

Gene Symbol
CT47A4
Entrez Gene ID
728075
Gene Name
cancer/testis antigen family 47, member A4
Gene Aliases
CT47.4
Chromosome Location
Chr. X: 120967886 - 120971306 on Build GRCh38
UniGene ID
Hs.729566
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080143.1 NP_001073612.1 3 1168

Gene Symbol
CT47A5
Entrez Gene ID
728072
Gene Name
cancer/testis antigen family 47, member A5
Gene Aliases
CT47.5
Chromosome Location
Chr. X: 120963026 - 120966446 on Build GRCh38
UniGene ID
Hs.729566
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080142.1 NP_001073611.1 3 1168

Gene Symbol
CT47A6
Entrez Gene ID
728062
Gene Name
cancer/testis antigen family 47, member A6
Gene Aliases
CT47.6
Chromosome Location
Chr. X: 120958165 - 120961588 on Build GRCh38
UniGene ID
Hs.723061
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080141.1 NP_001073610.1 3 1168

Gene Symbol
CT47A8
Entrez Gene ID
728049
Gene Name
cancer/testis antigen family 47, member A8
Gene Aliases
CT47.8
Chromosome Location
Chr. X: 120948422 - 120951842 on Build GRCh38
UniGene ID
Hs.729566
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080139.1 NP_001073608.1 3 1168

Gene Symbol
CT47A9
Entrez Gene ID
728042
Gene Name
cancer/testis antigen family 47, member A9
Gene Aliases
CT47.9
Chromosome Location
Chr. X: 120943561 - 120946981 on Build GRCh38
UniGene ID
Hs.729566
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080138.1 NP_001073607.1 3 1168

Gene Symbol
CT47B1
Entrez Gene ID
643311
Gene Name
cancer/testis antigen family 47, member B1
Gene Aliases
CT47.13, CT47A13
Chromosome Location
Chr. X: 120872554 - 120875925 on Build GRCh38
UniGene ID
Hs.729566
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145718.1 NP_001139190.1 3 1201
XM_017029734.1 XP_016885223.1 3 1160

View Full Product Details