Gene Transcripts
- Gene Symbol
- PLIN3
- Entrez Gene ID
- 10226
- Gene Name
- perilipin 3
- Gene Aliases
- M6PRBP1, PP17, TIP47
- Chromosome Location
- Chr. 19: 4838334 - 4867768 on Build GRCh38
- UniGene ID
- Hs.140452
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164189.1 |
NP_001157661.1
|
6, 7 |
1015 |
|
NM_001164194.1 |
NP_001157666.1
|
6, 7 |
979 |
|
NM_005817.4 |
NP_005808.3
|
6, 7 |
1015 |
|
GenBank mRNA |
AF051314.1 |
- |
2, 3 |
327 |
|
AF051315.1 |
- |
2, 3 |
303 |
|
AF055574.1 |
- |
6, 7 |
891 |
|
AF057140.1 |
- |
6, 7 |
912 |
|
AK122671.1 |
- |
6, 7 |
866 |
|
AK223054.1 |
- |
6, 7 |
945 |
|
AK225045.1 |
- |
6, 7 |
881 |
|
AK225400.1 |
- |
6, 7 |
896 |
|
AK226088.1 |
- |
6, 7 |
893 |
|
AK291104.1 |
- |
6, 7 |
972 |
|
AK312740.1 |
- |
6, 7 |
941 |
|
BC001590.2 |
- |
6, 7 |
888 |
2987965 |
BC005818.2 |
- |
6, 7 |
904 |
3833411 |
BC007566.1 |
- |
6, 7 |
904 |
3028104 |
BC019278.1 |
- |
6, 7 |
904 |
2905275 |
BQ063091.1 |
- |
|
869 |
|
BT007235.1 |
- |
5, 6 |
838 |
|
View Full Product Details