Gene Transcripts
- Gene Symbol
- CTAGE6
- Entrez Gene ID
- 340307
- Gene Name
- CTAGE family member 6
- Gene Aliases
- CTAGE6P
- Chromosome Location
- Chr. 7: 143755089 - 143757750 on Build GRCh38
- UniGene ID
- Hs.676333
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178561.4 |
NP_848656.2
|
1 |
2457 |
|
- Gene Symbol
- CTAGE15
- Entrez Gene ID
- 441294
- Gene Name
- CTAGE family member 15
- Gene Aliases
- CTAGE15P, cTAGE-15
- Chromosome Location
- Chr. 7: 143571801 - 143574387 on Build GRCh38
- UniGene ID
- Hs.450233
- Species
- Homo sapiens
View Full Product Details