Gene Transcripts

Gene Symbol
Fgd1
Entrez Gene ID
14163
Gene Name
FYVE, RhoGEF and PH domain containing 1
Gene Aliases
ZFYVE3
Chromosome Location
Chr. X: 151046118 - 151089686 on Build GRCm38
UniGene ID
Mm.475704
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008001.4 NP_032027.2 2 839
GenBank mRNA AK148151.1 - 2 840
BC011462.1 - 2 776 3674293
CN702035.1 - 2 329
U22325.1 - 2 438

View Full Product Details