Gene Transcripts

Gene Symbol
H2-DMb1
Entrez Gene ID
14999
Gene Name
histocompatibility 2, class II, locus Mb1
Gene Aliases
AI385589, H-2Mb1, H2-Mb1
Chromosome Location
Chr. 17: 34153191 - 34160229 on Build GRCm38
UniGene ID
Mm.460938
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010387.3 NP_034517.2 6 1240
GenBank mRNA AK032745.1 - 6 1241
BC002237.1 - 1201 3491702
BC003718.1 - 6 1180 3486844
X62743.1 - 1140

View Full Product Details