Gene Transcripts

Gene Symbol
Hba-x
Entrez Gene ID
15126
Gene Name
hemoglobin X, alpha-like embryonic chain in Hba complex
Gene Aliases
AI450015, Hbz
Chromosome Location
Chr. 11: 32276600 - 32278116 on Build GRCm38
UniGene ID
Mm.141758
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010405.4 NP_034535.1 3 449
GenBank mRNA AK077590.1 - 3 448
BC051988.1 - 3 419 5711898
M13125.1 - 2 257
M26898.1 - 3 407

View Full Product Details