Gene Transcripts

Gene Symbol
Foxf1
Entrez Gene ID
15227
Gene Name
forkhead box F1
Gene Aliases
AI450827, FREAC1, Foxf1a, Freac-1, HFH-8, Hfh8
Chromosome Location
Chr. 8: 121084386 - 121088154 on Build GRCm38
UniGene ID
Mm.33897
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010426.2 NP_034556.2 1 949
GenBank mRNA BC138805.1 - 1 949
BC138806.1 - 1 949 8861828

View Full Product Details