Gene Transcripts

Gene Symbol
LAMC3
Entrez Gene ID
10319
Gene Name
laminin subunit gamma 3
Gene Aliases
OCCM
Chromosome Location
Chr. 9: 131009117 - 131093059 on Build GRCh38
UniGene ID
Hs.201805
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006059.3 NP_006050.3 2 484
XM_006716921.2 XP_006716984.1 2 427
XM_011518121.1 XP_011516423.1 2 427
GenBank mRNA AF041835.1 - 2 483
AK075536.1 - 2 427

View Full Product Details